Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: FVDQLVEGLL GGLSHWDAEH FLFIAEHGYL YEHNFAFFPG FPLALLVGTE
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%
Glycosylphosphatidylinositol (GPI) is a complex glycolipid that anchors many proteins to the cell surface. The biosynthetic pathway of GPI is mediated by sequential addition of sugars and other components to phosphatidylinositol. PIGV adds the second mannose to the GPI core.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dol-P-Man dependent GPI mannosyltransferase; FLJ20477; GPI mannosyltransferase 2; GPI mannosyltransferase II; GPI-MT-II; phosphatidylinositol glycan anchor biosynthesis, class V; Phosphatidylinositol-glycan biosynthesis class V protein; PIG-V; Ybr004c homolog
Gene Aliases: GPI-MT-II; HPMRS1; PIG-V; PIGV
UniProt ID: (Human) Q9NUD9
Entrez Gene ID: (Human) 55650
Molecular Function:
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support