Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: DIPFEQDEEI LRGRLLFRRR VSPECQQLIR WCLSLRPSER PSLDQIAAHP WMLGADGGAP ESCDLRLCTL DPDDVASTTS SSESL
PIM3 serine/threonine kinase or PIM-3 oncogene belongs to a family of protooncogenes. PIM3 is expressed in hepatocellular carcinoma development and plays a main role in the proliferation of human hepatoma cell lines. PIM3 also expressed in human pancreatic cancer cells and phosphorylates BAD to block Bad-mediated apoptosis in human pancreatic cancer cell lines. PIM3 is widely expressed, with the exception of no expression observed in colon, thymus, and small intestine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: pim-3 oncogene; serine/threonine kinase Pim-3; Serine/threonine-protein kinase pim-3
Gene Aliases: pim-3; PIM3
UniProt ID: (Human) Q86V86
Entrez Gene ID: (Human) 415116
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support