Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: HSQSRRPSSW RQEKITRTKE EALELINGYI QKIKSGEEDF ESLASQFSDC SSAKARGDLG AFSRGQMQKP FEDASFALRT GEMSGPVFTD SGIHIILRTE
PIN1 (Peptidylprolyl Cis/Trans Isomerase, NIMA-Interacting 1) is a Protein Coding gene. Diseases associated with PIN1 include Meibomian Cyst and Viral Exanthem. Among its related pathways are Transport of the SLBP independent Mature mRNA and Cytokine Signaling in Immune system. Gene Ontology (GO) annotations related to this gene include peptidyl-prolyl cis-trans isomerase activity and mitogen-activated protein kinase binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 5.2.1.8; NIMA-interacting; Peptidyl-prolyl cis-trans isomerase NIMA-interacting 1; Peptidyl-prolyl cis-trans isomerase Pin1; PPIase Pin1; prolyl isomerase; protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1; Rotamase Pin1
Gene Aliases: DOD; PIN1; UBL5
UniProt ID: (Human) Q13526
Entrez Gene ID: (Human) 5300
Molecular Function: chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support