Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPPKGKSGS GKAGKGGAAS GSDSADKKAQ GPKGGGNAVK VRHILCEKHG KIMEAMEKLK SGMRFNEVAA QYSEDKARQG GDLGWMTRGS MVGPFQEAAF ALPVSGMDKP VFTDPPVKTK FGYHIIMVEG RK (10-140 aa encoded by BC005234)
This gene encodes a member of the parvulin subfamily of the peptidyl-prolyl cis/trans isomerase protein family. The encoded protein catalyzes the isomerization of peptidylprolyl bonds, and may play a role in the cell cycle, chromatin remodeling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eukaryotic parvulin homolog; hEPVH; hPar14; hPar17; MGC138486; Par14; Par17; parvulin; Parvulin-14; Parvulin-17; Peptidyl-prolyl cis-trans isomerase NIMA-interacting 4; Peptidyl-prolyl cis-trans isomerase Pin4; Peptidyl-prolyl cis/trans isomerase EPVH; peptidylprolyl cis/trans isomerase NIMA-interacting, 4; PPIase PIN4; protein (peptidylprolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin); Rotamase Pin4
Gene Aliases: 2410002I22Rik; EPVH; PAR14; PAR17; PIN4
UniProt ID: (Human) Q9Y237, (Mouse) Q9CWW6
Entrez Gene ID: (Human) 5303, (Rat) 684441, (Mouse) 69713
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support