Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Microtubule-binding protein essential for faithful chromosome segregation. Mediates TRF1 and TERT accumulation in nucleolus and enhances TRF1 binding to telomeres. Inhibits telomerase activity. May inhibit cell proliferation and act as tumor suppressor. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 67-11-3 protein; FLJ20565; hepatocellular carcinoma-related putative tumor suppressor; Liver-related putative tumor suppressor; LPTS1; MGC8850; PIN2 interacting protein 1; pin2-interact; PIN2-interacting protein 1; PIN2-interacting protein X1; PIN2/TERF1-interacting telomerase inhibitor 1; PIN2/TRF1-interacting protein; pinx1 {ECO:0000312|EMBL:ABO28828.1}; Protein 67-11-3; TRF1-interacting protein 1
Gene Aliases: 2210403I16Rik; 2610028A01Rik; 67-11-3; AU024023; LPTL; LPTS; LPTS1; PINX1; RGD1566025
UniProt ID: (Human) Q96BK5, (Rat) A4L691, (Mouse) Q9CZX5
Entrez Gene ID: (Human) 54984, (Rat) 305963, (Mouse) 72400
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support