Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSSNCTSTT AVAVAPLSAS KTKTKKKHFV CQKVKLFRAS EPILSVLMWG VNHTINELSN VPVPVMLMPD DFKAYSKIKV DNHLFNKENL PSRFKFKEYC PMVFRNLRER FGIDDQDYQN SVTRSAPINS DSQGRCGTRF LTTYDRRFVI KTVSSEDVAE MHNILKKYHQ FIVECHGNTL LPQFLGMYRL TVDGVETYMV VTRNVFSHRL TVHRKYDLKG STVAREASDK EKAKDLPTFK DNDFLNEGQK LHVGEESKKN FLEKLKRDVE EILVLSPGRR IA (1-281 aa encoded by BC027459)
The protein encoded by this gene catalyzes the phosphorylation of phosphatidylinositol-5-phosphate on the fourth hydroxyl of the myo-inositol ring to form phosphatidylinositol-5,4-bisphosphate. This gene is a member of the phosphatidylinositol-5-phosphate 4-kinase family. The encoded protein sequence does not show similarity to other kinases, but the protein does exhibit kinase activity. Additionally, the encoded protein interacts with p55 TNF receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-phosphatidylinositol 5-phosphate 4-kinase 2-beta; 1-phosphatidylinositol-4-phosphate kinase; 1-phosphatidylinositol-5-phosphate 4-kinase 2-beta; 1-phosphatidylinositol-5-phosphate 4-kinase B; Diphosphoinositide kinase 2-beta; Phosphatidylinositol 5-phosphate 4-kinase type II beta; Phosphatidylinositol 5-phosphate 4-kinase type-2 beta; phosphatidylinositol-4-phosphate 5-kinase, type II, beta; phosphatidylinositol-5-phosphate 4-kinase type-2 beta; phosphatidylinositol-5-phosphate 4-kinase, type II, beta; Phosphatidylinositol-phosphate kinase IIgamma; PI(5)P 4-kinase type II beta; PIP4KII-beta; PIPKIIgamma; PTDINS(4)P-5-kinase; PtdIns(5)P-4-kinase isoform 2-beta
Gene Aliases: AI848124; c11; PI5P4KB; PI5P4Kbeta; PIP4K2B; PIP5K2B; PIP5KIIB; PIP5KIIbeta; PIP5P4KB
UniProt ID: (Human) P78356, (Rat) O88377, (Mouse) Q80XI4
Entrez Gene ID: (Human) 8396, (Rat) 89812, (Mouse) 108083
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support