Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MASSSVPPA TVSAATAGPG PGFGFASKTK KKHFVQQKVK VFRAADPLVG VFLWGVAHSI NELSQVPPPV MLLPDDFKAS SKIKVNNHLF HRENLPSHFK FKEYCPQVFR NLRDRFGIDD QDYLVSLTRN PPSESEGSDG RFLISYDRTL VIKEVSSEDI ADMHSNLSNY HQYIVKCHGN TLLPQFLGMY RVSVDNEDSY MLVMRNMFSH RLPVHRKYDL KGSLVSREAS DKEKVKELPT LRDMDFLNKN QKVYIGEEEK KIFLEKLKRD VEFLVQLKIM DYSLLLGIHD IIRGSEPEEE APVREDESEV DGDCSLTGPP ALVGSYGTSP EGIGGYIHSH RPLGPGEFES FIDVYAIRSA EGAPQKEVYF MGLIDILTQY DAKKKAAHAA KTVKHGAGAE ISTVHPEQYA KRFLDFITNI FA (1-421 aa encoded by BC028596)
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: phosphatidylinositol 4-phosphate 5-kinase; Phosphatidylinositol 5-phosphate 4-kinase type II gamma; Phosphatidylinositol 5-phosphate 4-kinase type-2 gamma; phosphatidylinositol-4-phosphate 5-kinase, type II, gamma; phosphatidylinositol-5-phosphate 4-kinase type-2 gamma; phosphatidylinositol-5-phosphate 4-kinase, type II, gamma; PI(5)P 4-kinase type II gamma; PIP4KII-gamma
Gene Aliases: PIP4K2C; PIP5K2C
UniProt ID: (Human) Q8TBX8, (Mouse) Q91XU3, (Rat) O88370
Entrez Gene ID: (Human) 79837, (Mouse) 117150, (Rat) 140607
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support