Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: RPQEEPPAEE DLQQITVQVE PACSVEIVVP KEEDAGVEAS PAGASAAVEV ETASQASDEE GAPASQASDE EDAPATDIYF PTDERSWVYS PLHYSAQAPP ASDGESD
PIP5K1C lipid kinase or phosphatidylinositol-4-phosphate 5-kinase 1 gamma phosphorylates phosphatidylinositol-4-phosphate to form phosphatidylinositol 4,5-bisphosphate. PIP5K1C is found at synapses and has been found to play roles in endocytosis and cell migration. PIP5K1C directly associates with -beta-catenin and increases -beta-catenin activity downstream of growth factor stimulation. PIP5K1C expression and kinase activity enhance -beta-catenin phosphorylation on residues that promote nuclear importation and transcriptional activity. PIP5K1C deficiency impairs neutrophil recruitment as a result of defect in adhesion. PIP5K1C regulated the adhesion through facilitation of RhoA GTPase and integrin activation by chemo attractants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: diphosphoinositide kinase; Phosphatidylinositol 4-phosphate 5-kinase type-1 gamma; phosphatidylinositol-4-phosphate 5-kinase; phosphatidylinositol-4-phosphate 5-kinase, type I, gamma; PIP5K1gamma; PIP5KI gamma; PIP5KI-gamma; PIPKIg_v4; ptdIns(4)P-5-kinase 1 gamma; Type I phosphatidylinositol 4-phosphate 5-kinase gamma; type I PIP kinase
Gene Aliases: KIAA0589; LCCS3; PIP5K-GAMMA; PIP5K1-gamma; PIP5K1C; PIP5Kgamma
UniProt ID: (Human) O60331
Entrez Gene ID: (Human) 23396
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support