Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
This gene encodes a member of the cupin superfamily. The encoded protein is an Fe(II)-containing nuclear protein expressed in all tissues of the body and concentrated within dot-like subnuclear structures. Interactions with nuclear factor I/CCAAT box transcription factor as well as B cell lymphoma 3-encoded oncoprotein suggest the encoded protein may act as a transcriptional cofactor and be involved in the regulation of DNA transcription and replication. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: iron-binding nuclear protein; Pirin; pirin (iron-binding nuclear protein); pirin like; pirinin; Probable quercetin 2,3-dioxygenase PIR; Probable quercetinase
Gene Aliases: 2310042L19Rik; PIR; Pirnl
UniProt ID: (Human) O00625, (Rat) Q5M827, (Mouse) Q9D711
Entrez Gene ID: (Human) 8544, (Rat) 363465, (Mouse) 69656
Molecular Function:
transcription cofactor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support