Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Phosphatidylserine decarboxylases catalyze the formation of phosphatidylethanolamine (PE) by decarboxylation of phosphatidylserine (PS). Type I PSDs, such as PISD, are targeted to the inner mitochondrial membrane by an N-terminal targeting sequence. PISD also contains a conserved LGST motif that functions as an autocatalytic cleavage site where the proenzyme is split into mature alpha and beta subunits.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp566G2246; Phosphatidylserine decarboxylase proenzyme, mitochondrial
Gene Aliases: DJ858B16; dJ858B16.2; PISD; PSD; PSDC; PSSC
UniProt ID: (Human) Q9UG56
Entrez Gene ID: (Human) 23761
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support