Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
This Antibody was verified by Knockout to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 4
This gene encodes a cytoplasmic protein that catalyzes the transfer of phosphatidylinositol and phosphatidylcholine between membranes. This transfer activity is required for COPI complex-mediated retrograde transport from the Golgi apparatus to the endoplasmic reticulum. Alternative splicing of this gene results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Phosphatidylinositol transfer protein beta isoform; phosphatidylinositol transfer protein, beta; phosphotidylinositol transfer protein, beta; PI-TP beta; PI-TP-beta; PtdIns transfer protein beta
Gene Aliases: PI-TP-beta; PITPNB; PtdInsTP; VIB1B
UniProt ID: (Human) P48739
Entrez Gene ID: (Human) 23760
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support