Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MWRCGGRQGL CVLRRLSGGH AHHRAWRWNS NRACERALQY KLGDKIHGFT VNQVTSVPEL FLTAVKLTHD DTGARYLHLA REDTNNLFSV QFRTTPMDST GVPHILEHTV LCGSQKYPCR DPFFKMLNRS LSTFMNAFTA SDYTVYPFST QNPKDFQNLL SVYLDATFFP CLRELDFWQE GWRLEHENPS DPQTPLVFKG VVFNEMKGAF TDNERIFSQH LQNRLLPDHT YSVVSGGDPL CIPELTWEQL KQFHATHYHP SNARFFTYGN FPLEQHLKQI HEEALSKFQK IEPSTVVPAQ TPWDKPREFQ ITCGPDSFAT DPSKQTTVSV SFLLPDITDT FEAFTLSLLS SLLTSGPNSP FYKALIESGL GTDFSPDVGY NGYTREAYFS VGLQGIVEKD IETVRSLIDR TIDEVVEKGF EDDRIEALLH KIEIQMKHQS TSFGLMLTSY IASCWNHDGD PVELLKLGNQ LAKFRQCLQE NPKFLQEKVK QYFKNNQHKL TLSMRPDDKY HEKQAQVEAT KLKQKVEALS PGDRQQIYEK GLELRSQQSK PQDASCLPAL KVSDIEPTIP VTELDVVLTA GDIPVQYCAQ PTNGMVYFRA FSSLNTLPEE LRPYVPLFCS VLTKLGCGLL DYREQAQQIE LKTGGMSASP HVLPDDSHMD TYEQGVLFSS LCLDRNLPDM MQLWSEIFNN PCFEEEEHFK VLVKMTAQEL ANGIPDSGHL YASIRAGRTL TPAGDLQETF SGMDQVRLMK RIAEMTDIKP ILRKLPRIKK HLLNGDNMRC SVNATPQQMP QTEKAVEDFL RSIGRSKKER RPVRPHTVEK PVPSSSGGDA HVPHGSQVIR KLVMEPTFKP WQMKTHFLMP FPVNYVGECI RTVPYTDPDH ASFEILARLM TAKFLHTEIR EKGGAYGGGA KLSHNGIFTL YSYRDPNTIE TLQSFGKAVD WAKSGKFTQQ DIDEAKLSVF STVDAPVAPS DKGMDHFLYG LSDEMKQAHR EQLFAVSHDK LLAVSDRYLG TGKSTHGLAI LGPENPKIAK DPSWIIR
The protein encoded by this gene is an ATP-dependent metalloprotease that degrades post-cleavage mitochondrial transit peptides. The encoded protein binds zinc and can also degrade amyloid beta A4 protein, suggesting a possible role in Alzheimer's disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPreP; Metalloprotease 1; metalloprotease 1 (pitrilysin family); Pitrilysin metalloproteinase 1; PreP peptidasome; Presequence protease, mitochondrial
Gene Aliases: KIAA1104; MP1; PITRM1; PREP
UniProt ID: (Human) Q5JRX3
Entrez Gene ID: (Human) 10531
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support