Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 150 µL of MQ water. Centrifuge to remove any insoluble material.
Specificity of this antibody: PITX1.
PITX1 is a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. Members of this family are involved in organ development and left-right asymmetry. This protein acts as a transcriptional regulator involved in basal and hormone-regulated activity of prolactin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hindlimb-expressed homeobox protein backfoot; Homeobox protein P-OTX; Homeobox protein PITX1; OTX; Paired-like homeodomain transcription factor 1; Pituitary homeobox 1; Pituitary OTX-related factor
Gene Aliases: Bft; P-OTX; Pitx1; Potx; Ptx1
UniProt ID: (Mouse) P70314, (Rat) Q99NA7
Entrez Gene ID: (Mouse) 18740, (Rat) 113983
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support