Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of MQ water. Centrifuge to remove any insoluble material.
Specificity of this antibody: PITX2.
Pilx2 is a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. This protein acts as a transcription factor and regulates procollagen lysyl hydroxylase gene expression. It plays a role in the terminal differentiation of somatotroph and lactotroph cell phenotypes, is involved in the development of the eye, tooth and abdominal organs, and acts as a transcriptional regulator involved in basal and hormone-regulated activity of prolactin. Mutations in this protein are associated with Axenfeld-Rieger syndrome, iridogoniodysgenesis syndrome, and sporadic cases of Peters anomaly. A similar protein in other vertebrates is involved in the determination of left-right asymmetry during development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ALL1-responsive protein ARP1; Bicoid-related homeobox protein 1; BRX1 homeoprotein; Homeobox protein PITX2; Orthodenticle-like homeobox 2; Paired-like homeodomain transcription factor 2; paired-like homeodomain transcription factor Munc 30; Pituitary homeobox 2; S a 1 glycoprotein; S a1glycoprotein; S alpha 1 glycoprotein; S α 1 glycoprotein; S α1glycoprotein; Solurshin
Gene Aliases: 9430085M16Rik; Arp1; Brx1; Brx1b; Munc30; Otlx2; Pitx2; Ptx2; Rgs; Rieg
UniProt ID: (Mouse) P97474
Entrez Gene ID: (Mouse) 18741
Molecular Function:
homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support