Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNCMKGPLHL EHRAAGTKLS AVSSSSCHHP QPLAMASVLA PGQPRSLDSS KHRLEVHTIS DTSSPEAAEK DKSQQGKNED VGAEDPSKKK RQRRQRTHFT SQQLQELEAT FQRNRYPDMS TREEIAVWTN LTEARVRVWF KNRRAKWRKR ERNQQAELCK NGFGPQFNGL MQPYDDMYPG YSYNNWAAKG LTSASLSTKS FPFFNSMNVN PLSSQSMFSP PNSISSMSMS SSMVPSAVTG VPGSSLNSLN NLNNLSSPSL NSAVPTPACP YAPPTPPYVY RDTCNSSLAS LRLKAKQHSS FGYASVQNPA SNLSACQYAV DRPV
Pilx2 is a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. This protein acts as a transcription factor and regulates procollagen lysyl hydroxylase gene expression. It plays a role in the terminal differentiation of somatotroph and lactotroph cell phenotypes, is involved in the development of the eye, tooth and abdominal organs, and acts as a transcriptional regulator involved in basal and hormone-regulated activity of prolactin. Mutations in this protein are associated with Axenfeld-Rieger syndrome, iridogoniodysgenesis syndrome, and sporadic cases of Peters anomaly. A similar protein in other vertebrates is involved in the determination of left-right asymmetry during development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: all1-responsive gene 1; ALL1-responsive protein ARP1; Homeobox protein PITX2; paired-like homeodomain 2; Paired-like homeodomain transcription factor 2; Pituitary homeobox 2; RIEG bicoid-related homeobox transcription factor; rieg bicoid-related homeobox transcription factor 1; S a 1 glycoprotein; S a1glycoprotein; S alpha 1 glycoprotein; S α 1 glycoprotein; S α1glycoprotein; Solurshin
Gene Aliases: ARP1; Brx1; IDG2; IGDS; IGDS2; IHG2; IRID2; Otlx2; PITX2; PTX2; RGS; RIEG; RIEG1; RS
UniProt ID: (Human) Q99697
Entrez Gene ID: (Human) 5308
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support