Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Pilx2 is a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. This protein acts as a transcription factor and regulates procollagen lysyl hydroxylase gene expression. It plays a role in the terminal differentiation of somatotroph and lactotroph cell phenotypes, is involved in the development of the eye, tooth and abdominal organs, and acts as a transcriptional regulator involved in basal and hormone-regulated activity of prolactin. Mutations in this protein are associated with Axenfeld-Rieger syndrome, iridogoniodysgenesis syndrome, and sporadic cases of Peters anomaly. A similar protein in other vertebrates is involved in the determination of left-right asymmetry during development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: all1-responsive gene 1; ALL1-responsive protein ARP1; Bicoid-related homeobox protein 1; BRX1 homeoprotein; Homeobox protein PITX2; homeodomain transcription factor 2; Orthodenticle-like homeobox 2; paired-like homeodomain 2; Paired-like homeodomain transcription factor 2; paired-like homeodomain transcription factor Munc 30; Pituitary homeobox 2; RIEG bicoid-related homeobox transcription factor; rieg bicoid-related homeobox transcription factor 1; rPtx2; S a 1 glycoprotein; S a1glycoprotein; S alpha 1 glycoprotein; S α 1 glycoprotein; S α1glycoprotein; Solurshin
Gene Aliases: 9430085M16Rik; ARP1; Brx1; Brx1b; IDG2; IGDS; IGDS2; IHG2; IRID2; Munc30; Otlx2; Pitx-2; PITX2; Pitx2c; PTX2; RGS; RIEG; RIEG1; RS
UniProt ID: (Human) Q99697, (Rat) Q9R0W1, (Mouse) P97474
Entrez Gene ID: (Human) 5308, (Rat) 54284, (Mouse) 18741
Molecular Function:
homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support