Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
cAMP is a signaling molecule important for a variety of cellular functions. cAMP exerts its effects by activating the cAMP-dependent protein kinase (AMPK), which transduces the signal through phosphorylation of different target proteins. The inactive holoenzyme of AMPK is a tetramer composed of two regulatory and two catalytic subunits. cAMP causes the dissociation of the inactive holoenzyme into a dimer of regulatory subunits bound to four cAMP and two free monomeric catalytic subunits. Four different regulatory subunits and three catalytic subunits of AMPK have been identified in humans. PRKACB is a member of the Ser/Thr protein kinase family and is a catalytic subunit of AMPK.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cAMP-dependent protein kinase C beta; cAMP-dependent protein kinase catalytic beta subunit isoform 4ab; cAMP-dependent protein kinase catalytic subunit alpha; cAMP-dependent protein kinase catalytic subunit alpha, isoform 1; cAMP-dependent protein kinase catalytic subunit beta; cAMP-dependent protein kinase catalytic subunit C alpha; PKA C-alpha; PKA C-beta; protein kinase A; protein kinase A catalytic subunit; protein kinase A catalytic subunit beta; protein kinase beta-catalytic subunit; protein kinase, cAMP-dependent, alpha catalytic subunit; protein kinase, cAMP-dependent, beta catalytic subunit; protein kinase, cAMP-dependent, catalytic, alpha; protein kinase, cAMP-dependent, catalytic, beta; sperm cAMP-dependent protein kinase catalytic subunit
Gene Aliases: Cs-PKA; PKA C-beta; PKACA; PKACB; PKCA1; PKCD; PPNAD4; PRKACA; PRKACB
UniProt ID: (Human) P17612, (Bovine) P00517, (Human) P22694, (Bovine) P05131, (Mouse) P05132, (Rat) P68182, (Mouse) P68181
Entrez Gene ID: (Human) 5566, (Bovine) 282322, (Human) 5567, (Bovine) 282323, (Rat) 25636, (Mouse) 18747, (Rat) 293508, (Mouse) 18749
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support