Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: PVSKTEKTNF KTLSSMEDFW KFTEGSLLDG LYWKMQPSNQ TEADNRSFIF YENLLLGVPR IRQLRVRNGS CSIPQDLRDE IKECYDVYSV SSEDRAPFGP
This gene encodes a member of the polycystin protein family. The encoded protein contains multiple transmembrane domains, and cytoplasmic N- and C-termini. The protein may be an integral membrane protein involved in cell-cell/matrix interactions. The encoded protein may function in renal tubular development, morphology, and function, and may modulate intracellular calcium homoeostasis and other signal transduction pathways. This protein interacts with polycystin 1 to produce cation-permeable currents. Mutations in this gene have been associated with autosomal dominant polycystic kidney disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A MGC138466; Autosomal dominant polycystic kidney disease type II protein; MGC138468; PC2; polycystic kidney disease 2 (autosomal dominant); Polycystic kidney disease 2 protein; Polycystin-2; Polycystwin; R48321; Transient receptor potential cation channel subfamily P member 2; transient receptor potential cation channel, subfamily P, member 2; TRPP1
Gene Aliases: APKD2; Pc-2; PC2; PKD2; PKD4; TRPP2
UniProt ID: (Human) Q13563
Entrez Gene ID: (Human) 5311
Molecular Function: ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support