Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NDTYSEVKE ELAGQKDELQ LSDLLKQGYN KTLLRLRLRK ERVSDVQKVL QGGEQEIQFE DFTNTLRELG HAEHEITELT ATFTKFDRDG NRILDEKEQE KMRQDLEEER VALNTEIEKL GRSIVSSPQG KSGPEAARAG GWVSGEEFYM LTRRVLQLET VLEGVVSQID AVGSKLKMLE RKGWLAPSPG VKEQAIWKHP QPAPAVTPDP WGVQGGQESE VPYKREEEAL EERRLSRGEI PTLQRS (561-805 aa encoded by BC025665)
This gene encodes a member of the polycystin protein family. The encoded protein contains multiple transmembrane domains, and cytoplasmic N- and C-termini. The protein may be an integral membrane protein involved in cell-cell/matrix interactions. This protein functions as a calcium-regulated nonselective cation channel. Alternative splice variants have been described but their full length sequences have not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PK2L1; Polycystic kidney disease 2-like 1 protein; Polycystin-2 homolog; Polycystin-2-like protein 1; Polycystin-2L1; Polycystin-L; Polycystin-L1; transient receptor potential cation channel, subfamily P, member 3
Gene Aliases: B830002B15; BC046386; PCL; PKD2L; PKD2L1; PKDL; TRPP3
UniProt ID: (Human) Q9P0L9, (Mouse) A2A259
Entrez Gene ID: (Human) 9033, (Rat) 293937, (Mouse) 329064
Molecular Function:
ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support