Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: KWYNNQSLGH GSHSFIYYEN MLLGVPRLRQ LKVRNDSCVV HEDFREDILS CYDVYSPDKE EQLPFGPFNG TAWTYHSQDE LGGFSHWGRL TSYSGGGYYL
This gene encodes a member of the polycystin protein family. The encoded protein contains multiple transmembrane domains, and cytoplasmic N- and C-termini. The protein may be an integral membrane protein involved in cell-cell/matrix interactions. This protein functions as a calcium-regulated nonselective cation channel. Alternative splice variants have been described but their full length sequences have not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PK2L1; Polycystic kidney disease 2-like 1 protein; Polycystin-2 homolog; Polycystin-2-like protein 1; Polycystin-2L1; Polycystin-L; Polycystin-L1; transient receptor potential cation channel, subfamily P, member 3
Gene Aliases: PCL; PKD2L; PKD2L1; PKDL; TRPP3
UniProt ID: (Human) Q9P0L9
Entrez Gene ID: (Human) 9033, (Rat) 293937
Molecular Function:
ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support