Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MASDAVQSEP RSWSLLEQLG LAGADLAAPG VQQQLELERE RLRREIRKEL KLKEGAENLR RATTDLGRSL GPVELLLRGS SRRLDLLHQQ LQELHAHVVL PDPAATHDGP QSPGAGGPTC SATNLSRVAG LEKQLAIELK VKQGAENMIQ TYSNGSTKDR KLLLTAQQML QDSKTKIDII RMQLRRALQA GQLENQAAPD DTQGSPDLGA VELRIEELRH HFRVEHAVAE GAKNVLRLLS AAKAPDRKAV SEAQEKLTES NQKLGLLREA LERRLGELPA DHPKGRLLRE ELAAASSAAF STRLAGPFPA THYSTLCKPA PLTGTLEVRV VGCRDLPETI PWNPTPSMGG PGTPDSRPPF LSRPARGLYS RSGSLSGRSS LKAEAENTSE VSTVLKLDNT VVGQTSWKPC GPNAWDQSFT LELERARELE LAVFWRDQRG LCALKFLKLE DFLDNERHEV QLDMEPQGCL VAEVTFRNPV IERIPRLRRQ KKIFSKQQGK AFQRARQMNI DVATWVRLLR RLIPNATGTG TFSPGASPGS EARTTGDISV EKLNLGTDSD SSPQKSSRDP PSSPSSLSSP IQESTAPELP SETQETPGPA LCSPLRKSPL TLEDFKFLAV LGRGHFGKVL LSEFRPSGEL FAIKALKKGD IVARDEVESL MCEKRILAAV TSAGHPFLVN LFGCFQTPEH VCFVMEYSAG GDLMLHIHSD VFSEPRAIFY SACVVLGLQF LHEHKIVYRD LKLDNLLLDT EGYVKIADFG LCKEGMGYGD RTSTFCGTPE FLAPEVLTDT SYTRAVDWWG LGVLLYEMLV GESPFPGDDE EEVFDSIVND EVRYPRFLSA EAIGIMRRLL RRNPERRLGS SERDAEDVKK QPFFRTLGWE ALLARRLPPP FVPTLSGRTD ISNFDEEFTG EAPTLSPPRD ARPLTAAEQA AFLDFDFVAG GC
PKN1 (PRK1) belongs to the protein kinase C superfamily. This kinase is activated by the Rho family of small G proteins and may mediate the Rho-dependent signaling pathway. This kinase can also be activated by phospholipids. The 3-phosphoinositide dependent protein kinase-1 (PDPK1/PDK1) is reported to phosphorylate this kinase, which may mediate insulin signals to the actin cytoskeleton. The proteolytic activation of this kinase by caspase-3 or related proteases during apoptosis suggests its role in signal transduction related to apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been observed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PAK-1; Protease-activated kinase 1; Protein kinase C-like 1; Protein kinase C-like PKN; protein kinase C-related kinase 1; Protein kinase PKN-alpha; Protein-kinase C-related kinase 1; serine-threonine kinase N; Serine-threonine protein kinase N; serine/threonine protein kinase N; Serine/threonine-protein kinase N1
Gene Aliases: DBK; PAK-1; PAK1; PKN; PKN-ALPHA; PKN1; PRK1; PRKCL1
UniProt ID: (Human) Q16512
Entrez Gene ID: (Human) 5585
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support