Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QEQAQTTDW RATLKTIRNG VHKIDTYLNA ALDLLGGEDG LCQYKCSDGS KPFPRYGYKP SPPNGCGSPL FGVHLNIGIP SLTKCCNQHD RCYETCGKSK NDCDEEFQYC LSKICRDVQK TLGLTQHVQA CETTVELLFD SVIHLGCKPY LDSQRAACRC HYEEKTDL (23-189 aa encoded by BC017218 )
Secreted phospholipase A2 (sPLA2) enzymes liberate arachidonic acid from phospholipids for production of eicosanoids and exert a variety of physiologic and pathologic effects. Group XII sPLA2s, such as PLA2G12A, have relatively low specific activity and are structurally and functionally distinct from other sPLA2s.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: group XII secreted phospholipase A2; group XII-1 phospholipase A2; group XIIA secreted phospholipase A2; Group XIIA secretory phospholipase A2; GXII sPLA2; Phosphatidylcholine 2-acylhydrolase 12A; phospholipase A2, group XII; phospholipase A2, group XIIA; sPLA2-XII
Gene Aliases: 2310004B05Rik; FKSG38; GXII; GXII-1; GXII-1-PLA2; mGXII-1; PLA2G12; PLA2G12A; ROSSY; UNQ2519/PRO6012
UniProt ID: (Human) Q9BZM1, (Mouse) Q9EPR2
Entrez Gene ID: (Human) 81579, (Rat) 362039, (Mouse) 66350
Molecular Function:
phospholipase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support