Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PLA2G4B transcribes naturally-occurring mRNAs that are co-transcribed products of the neighboring JMJD7 and PLA2G4B genes. Incompletely processed read-through transcripts from these two loci are abundantly expressed in most tissues, but the function of the predicted protein product has not yet been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cPLA2-beta; Cytosolic phospholipase A2 beta; Lysophospholipase A1 group IVB; Phospholipase A2 group IVB; phospholipase A2, group IVB (cytosolic)
Gene Aliases: cPLA2-beta; HsT16992; PLA2G4B
UniProt ID: (Human) P0C869
Entrez Gene ID: (Human) 100137049
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support