Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KDDVSNQDF REWFEFSPYE VGLQKYGAFI PSELFGSEFF MGRLVKRIPE SRICYMLGLW SSIFSLNLLD AWNLSHTSEE FFHRWTREKV QDIEDEPILP EIPKCDANIL ETTVVIPGSW LSNSFREILT HRSFVSEFHN FLSGLQLHTN YLQNGQFSRW KDTVLDGFPN QLTESANHLC LLDTAFFVNS SYPPLLRPER KADLIIHLNY CAGSQTKPLK QTCEYCTVQN IPFPKYELPD ENENLKECYL MENPQEPDAP IVTFFPLIND TFRKYKAPGV ERSPEELEQG QVDIYGPKTP YATKELTYTE ATFDKLVKLS EYNILNNKDT LLQALRLAVE KKKRLKGQCP S (507-856 aa encoded by BC101584)
This gene encodes a member of the cytosolic phospholipase A2 group IV family. Members of this family are involved in regulation of membrane tubule-mediated transport. The enzyme encoded by this member of the family plays a role in trafficking through the clathrin-independent endocytic pathway. The enzyme regulates the recycling process via formation of tubules that transport internalized clathrin-independent cargo proteins back to the cell surface.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcium-dependent N-acyltransferase; cPLA2-epsilon; Cytosolic phospholipase A2 epsilon; EC 3.1.1.4; Phospholipase A2 group IVE; phospholipase A2, group 4E; phospholipase A2, group IVE
Gene Aliases: 2310026J01Rik; C230096D22; Pla2epsilon; PLA2G4E; RGD1310595
UniProt ID: (Human) Q3MJ16, (Mouse) Q50L42
Entrez Gene ID: (Human) 123745, (Rat) 296091, (Mouse) 329502
Molecular Function:
phospholipase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support