Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GAGRYVPGS ASMGTTMAGV DPFTGNSAYR SAASKTMNIY FPKKEAVTFD QANPTQILGK LKELNGTAPE EKKLTEDDLI LLEKILSLIC NSSSEKPTVQ QLQILWKAIN CPEDIVFPAL DILRLSIKHP SVNENFCNEK EGAQFSSHLI NLLNPKGKPA NQLLALRTFC NCFVGQAGQK LMMSQRESLM SHAIELKSGS NKNIHIALAT LALNYSVCFH KDHNIEGKAQ CLSLISTILE VVQDLEATFR LLVALGTLIS DDSNAVQLAK SLGVDSQIKK YSSVSEPAKV SECCRFILNL L (496-795 aa encoded by BC032551)
Phospholipase A-2-activating protein (PLAA), also named PLAP, can interact with ubiquitin and is involved in the maintenance of ubiquitin levels. PLAA contains multiple consensus domains including WD repeats, PFU domain and PUL domain. Although the function of PLAA remains unclear, ubiquitin binding of PLAA might be the central role that connects ubiquitination and degradation in ERAD. Recent finding revealed that PLAA served as a novel nociceptive mediator after incision, and its expression level is regulated by miR-203.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DOA1 homolog; Phospholipase A-2-activating protein; phospholipase A2-activating protein; PLA2P
Gene Aliases: 2410007N06; AI536418; AU018445; AW208417; D4Ertd618e; DOA1; PLA2P; PLAA; PLAP; Ufd3
UniProt ID: (Human) Q9Y263, (Mouse) P27612, (Rat) P54319
Entrez Gene ID: (Human) 9373, (Mouse) 18786, (Rat) 116645
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support