Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PLBD2 (Phospholipase B Domain Containing 2) is a Protein Coding gene. Diseases associated with PLBD2 include Gallbladder Papillomatosis and Gm1-Gangliosidosis, Type Iii. Gene Ontology (GO) annotations related to this gene include hydrolase activity. An important paralog of this gene is PLBD1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 66.3 kDa protein; 76 kDa protein; LAMA-like protein 2; Lamina ancestor homolog 2; p76; Phospholipase B domain-containing protein 2; Putative phospholipase B-like 2
Gene Aliases: 1300012G16Rik; AU019810; P76; Plbd2
UniProt ID: (Mouse) Q8C0M4
Entrez Gene ID: (Mouse) 71772
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support