Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AQAGQETCQ DTQSQQLGSQ PSSNPTPSPL DASPRRPPGP TTSPASTSLS SPGQRDDLIA SILSEVAPTP LDELRGHKAL VKLRSRQERD LRELRKKHQR KAVTLTRRLL DGLAQAQAEG RCRLRPGALG GAADVEDTKE GEDEAKRYQE FQNRQVQSLL ELREAQVDAE AQRRLEHLRQ ALQRLREVVL DANTTQFKRL KEMNEREKKE LQKILDRKRH NSISEAKMRD KHKKEAELTE INRRHITESV NSIRRLEEAQ KQRHDRLVAG QQQVLQQLAE EEPKLLAQLA QECQEQRARL PQEIRRSLLG EMPEGLGDGP LVACASNGHA PGSSGHLSGA DSESQEENTQ L (885-1234 aa encoded by BC142681)
PLCB3 encodes a member of the phosphoinositide phospholipase C beta enzyme family that catalyze the production of the secondary messengers diacylglycerol and inositol 1,4,5-triphosphate from phosphatidylinositol in G-protein-linked receptor-mediated signal transduction. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-3; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-3; EC 3.1.4.11; Phosphoinositide phospholipase C-beta-3; phosphoinositide-specific phospholipase c; phospholipase C beta 3; phospholipase C, beta 3 (phosphatidylinositol-specific); Phospholipase C-beta-3; PLC beta 3; PLC-beta-3; PLC-beta3; PLCbeta3
Gene Aliases: mKIAA4098; PLCB3
UniProt ID: (Human) Q01970, (Mouse) P51432
Entrez Gene ID: (Human) 5331, (Rat) 29322, (Mouse) 18797
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support