Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MASLLQDQLT TDQDLLLMQE GMPMRKVRSK SWKKLRYFRL QNDGMTVWHA RQARGSAKPS FSISDVETIR NGHDSELLRS LAEELPLEQG FTIVFHGRRS NLDLMANSVE EAQIWMRGLQ LLVDLVTSMD HQERLDQWLS DWFQRGDKNQ DGKMSFQEVQ RLLHLMNVEM DQEYAFSLFQ AADTSQSGTL EGEEFVQFYK ALTKRAEVQE LFESFSADGQ KLTLLEFLDF LQEEQKERDC TSELALELID RYEPSDSGKL RHVLSMDGFL SYLCSKDGDI FNPACLPIYQ
This gene encodes a member of the delta class of phospholipase C enzymes. Phospholipase C enzymes play a critical role in many cellular processes by hydrolyzing phosphatidylinositol 4,5-bisphosphate into two intracellular second messengers, inositol 1,4,5-trisphosphate and diacylglycerol. Expression of this gene may be a marker for cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase delta-4; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-4; hPLCD4; phosphoinositide phospholipase C delta-4; Phosphoinositide phospholipase C-delta-4; phospholipase C, delta 4; Phospholipase C-delta-4; PLC delta4; PLC-delta-4
Gene Aliases: PLCD4
UniProt ID: (Human) Q9BRC7, (Rat) Q62711
Entrez Gene ID: (Human) 84812, (Rat) 140693
Molecular Function:
phospholipase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support