Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: RYLISYGKHT LDMLESSQDN MRTSWVSQMF SEIDVDNLGH ITLCNAVQCI RNLNPGLKTS KIELKFKELH KSKDKAGTEV TKEEFIEVFH
May play an role in the regulation of Ins(1,4,5)P3 around the endoplasmic reticulum.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Inactive phospholipase C-like protein 2; phospholipase C; phospholipase C, epsilon 2; Phospholipase C-epsilon-2; phospholipase C-L2; phospholipase C-like 2; PLC-epsilon-2; PLC-L(2); PLC-L2
Gene Aliases: KIAA1092; PLCE2; PLCL2
UniProt ID: (Human) Q9UPR0
Entrez Gene ID: (Human) 23228
Molecular Function:
phospholipase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support