Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEPKRIREG YLVKKGSVFN TWKPMWVVLL EDGIEFYKKK SDNSPKGMIP LKGSTLTSPC QDFGKRMFVF KITTTKQQDH FFQAAFLEER DAWVRDIKKA IKCIEGGQKF ARKSTRRSIR LPETIDLGAL YLSMKDTEKG IKELNLEKDK KIFNHCFTGN CVIDWLVSNQ SVRNRQEGLM IASSLLNEGY LQPAGDMSKS AVDGTAENPF LDNPDAFYYF PDSGFFCEEN SSDDDVILKE EFRGVIIKQG CLLKQGHRRK NWKVRKFILR EDPAYLHYYD PAGAEDPLGA IHLRGCVVTS VESNSNGRKS EEENLFEIIT ADEVHYFLQA ATPKERTEWI KAIQMASRTG K (1-350 aa encoded by BC018549)
Major protein kinase C substrate of platelets.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ27168; p47; Platelet 47 kDa protein; Pleckstrin
Gene Aliases: 2010300B13Rik; P47; PLEK
UniProt ID: (Human) P08567, (Mouse) Q9JHK5, (Rat) Q4KM33
Entrez Gene ID: (Human) 5341, (Mouse) 56193, (Rat) 364206
Molecular Function:
cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support