Sequence of this protein is as follows: MALDGIRMPD GCYADGTWEL SVHVTDLNRD VTLRVTGEVH IGGVMLKLVE KLDVKKDWSD HALWWEKKRT WLLKTHWTLD KYGIQADAKL QFTPQHKLLR LQLPNMKYVK VKVNFSDRVF KAVSDICKTF NIRHPEELSL LKKPRDPTKK KKKKLDDQSE DEALELEGPL ITPGSGSIYS SPGLYSKTMT PTYDAHDGSP LSPTSAWFGD SALSEGNPGI LAVSQPITSP EILAKMFKPQ ALLDKAKINQ GWLDSSRSLM EQDVKENEAL LLRFKYYSFF DLNPKYDAIR INQLYEQAKW AILLEEIECT EEEMMMFAAL QYHINKLSIM TSENHLNNSD KEVDEVDAAL SDLEITLEGG KTSTILGDIT SIPELADYIK VFKPKKLTLK GYKQYWCTFK DTSISCYKSK EESSGTPAHQ MNLRGCEVTP DVNISGQKFN IKLLIPVAEG MNEIWLRCDN EKQYAHWMAA CRLASKGKTM ADSSYNLEVQ NILSSLKMQH LNPDPQLIPE QITTDITPEC LVSPRYLKKY KNKQITARIL EAHQNVAQMS LIEAKMRFIQ AWQSLPEFGI THFIARFQGG KKEELIGIAY NRLIRMDAST GDAIKTWRFS NMKQWNVNWE IKMVTVEFAD EVRLSFICTE VDCKVVHEFI GGYIFLSTRA KDQNESLDEE MFYKLTSGWV
Scaffolding protein that enhances integrin activation mediated by TLN1 and/or TLN2, but activates integrins only weakly by itself. Binds to membranes enriched in phosphoinositides. Enhances integrin-mediated cell adhesion onto the extracellular matrix and cell spreading; this requires both its ability to interact with integrins and with phospholipid membranes. Required for the assembly of focal adhesions. Participates in the connection between extracellular matrix adhesion sites and the actin cytoskeleton and also in the orchestration of actin assembly and cell shape modulation. Recruits FBLIM1 to focal adhesions. Plays a role in the TGFB1 and integrin signaling pathways. Stabilizes active CTNNB1 and plays a role in the regulation of transcription mediated by CTNNB1 and TCF7L2/TCF4 and in Wnt signaling. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fermitin family homolog 2; kindlin 2; Kindlin-2; MIG-2; mitogen inducible gene 2 protein; Mitogen-inducible gene 2 protein; PH domain-containing family C member 1; pleckstrin homology domain containing, family C (with FERM domain) member 1; pleckstrin homology domain containing, family C member 1; Pleckstrin homology domain-containing family C member 1
Gene Aliases: FERMT2; KIND2; mig-2; MIG2; PLEKHC1; UNC112; UNC112B
UniProt ID: (Human) Q96AC1
Entrez Gene ID: (Human) 10979
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support