Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MSADGAEADG STQVTVEEPV QQPSVVDRVA SMPLISSTCD MVSAAYASTK ESYPHVKTVC DAAEKGVRTL TAAAVSGAQP ILSKLEPQIA SASEYAHRGL DKLEENLPIL QQPTEKVLAD TKELVSSKVS GAQEMVSSAK DTVATQLSEA VDATRGAVQS GVDKTKSVVT GGVQSVMGSR LGQMVLSGVD TVLGKSEEWA DNHLPLTDAE LARIATSLDG FDVASVQQQR QEQSYFVRLG SLSERLRQHA YEHSLGKLRA TKQRAQEALL QLSQALSLME TVKQGVDQKL VEGQEKLHQM WLSWNQKQLQ GPEKEPPKPE QVESRALTMF RDIAQQLQAT CTSLGSSIQG LPTNVKDQVQ QARRQVEDLQ ATFSSIHSFQ DLSSSILAQS RERVASAREA LDHMVEYVAQ NTPVTWLVGP FAPGITEKAP EEKK
TIP47 has been described as cargo protein involved in the trafficking of the mannose-6-phosphate receptor between endosomes and the Golgi complex. Mannose 6-phophate receptors (MPRs) deliver lysosomal hydrolase from the Golgi to endosomes and then return to the Golgi complex. The protein encoded by this gene interacts with the cytoplasmic domains of both cation-independent and cation-dependent MPRs and is required for endosome to Golgi transport. This protein also binds directly to the GTPase RAB9, a member of the RAS oncogene family. The interaction with RAB9 has been shown to increase the affinity of this protein for its cargo. TIP47 has been localized in milk fat globule membranes of human and bovine origin. It has been described as a placental protein, as well. Increased amounts of TIP47 are secreted into circulation of cervix carcinoma patients. Therefore, this protein is probably significant as an oncodevelopmental marker.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 47 kDa mannose 6-phosphate receptor-binding protein; 47 kDa MPR-binding protein; Cargo selection protein TIP47; Mannose-6-phosphate receptor-binding protein 1; MGC11117; MGC2012; Perilipin-3; Placental protein 17; PP17; tail-interacting protein, 47 kD; testicular tissue protein Li 114
Gene Aliases: M6PRBP1; PLIN3; PP17; TIP47
UniProt ID: (Human) O60664
Entrez Gene ID: (Human) 10226
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support