Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NRKIIAPLV TRHGKLWSNF WGALSPDGYY ARSEDYVDIV QGNRVGVWNV PYMANVYLIK GKTLRSEMNE RNYFVRDKLD PDMALCRNAR EMTLQREKDS PTPETFQMLS PPKGVFMYIS NRHEFGRLLS TANYNTSHYN NDLWQIFENP VDWKEKYINR DYSKIFTENI VEQPCPDVFW FPIFSEKACD ELVEEMEHYG KWSGGKHHDS RISGGYENVP TDDIHMKQVD LENVWLHFIR EFIAPVTLKV FAGYYTKGFA LLNFVVKYSP ERQRSLRPHH DASTFTINIA LNNVGEDFQG GGCKFLRYNC SIESPRKGWS FMHPGRLTHL HEGLPVKNGT RYIAVSFIDP (410-758 aa encoded by BC037169)
The protein encoded by this gene is a membrane-bound homodimeric enzyme that is localized to the cisternae of the rough endoplasmic reticulum. The enzyme (cofactors iron and ascorbate) catalyzes the hydroxylation of lysyl residues in collagen-like peptides. The resultant hydroxylysyl groups are attachment sites for carbohydrates in collagen and thus are critical for the stability of intermolecular crosslinks. Some patients with Ehlers-Danlos syndrome type VIB have deficiencies in lysyl hydroxylase activity. Mutations in the coding region of this gene are associated with Bruck syndrome. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LH2; lysine hydroxylase 2; lysyl hydroxlase 2; Lysyl hydroxylase 2; procollagen-lysine 5-dioxygenase; procollagen-lysine, 2-oxoglutarate 5-dioxygenase 2; Procollagen-lysine,2-oxoglutarate 5-dioxygenase 2; telopeptide lysyl hydroxylase
Gene Aliases: BRKS2; LH2; PLOD2; TLH
UniProt ID: (Human) O00469
Entrez Gene ID: (Human) 5352
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support