Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: HEPHIADSW PQLQDHFSAV KLVGPEEALS PGEARDMAMD LCRQDPECEF YFSLDADAVL TNLQTLRILI EENRKVIAPM LSRHGKLWSN FWGALSPDEY YARSEDYVEL VQRKRVGVWN VPYISQAYVI RGDTLRMELP QRDVFSGSDT DPDMAFCKSF RDKGIFLHLS NQHEFGRLLA TSRYDTEHLH PDLWQIFDNP VDWKEQYIHE NYSRALEGEG IVEQPCPDVY WFPLLSEQMC DELVAEMEHY GQWSGGRHED SRLAGGYENV PTVDIHMKQV GYEDQWLQLL RTYVGPMTES LFPGYHTKAR AVMNFVVRYR PDEQPSLRPH HDSSTFTLNV ALNHKGLDYE GGGCRFLRYD CVISSPRKGW ALLHPGRLTH YHEGLPTTWG TRYIMVSFVD P (339-738 aa encoded by BC011674)
Forms hydroxylysine residues in -Xaa-Lys-Gly- sequences in collagens. These hydroxylysines serve as sites of attachment for carbohydrate units and are essential for the stability of the intermolecular collagen cross-links.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: lysine hydroxylase 3; lysyl hydroxlase 3; lysyl hydroxylase 3; Multifunctional procollagen lysine hydroxylase and glycosyltransferase LH3; procollagen-lysine, 2-oxoglutarate 5-dioxygenase 3
Gene Aliases: LH3; PLOD3
UniProt ID: (Human) O60568
Entrez Gene ID: (Human) 8985
Molecular Function:
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support