Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MADSERLSA PGCWAACTNF SRTRKGILLF AEIILCLVIL ICFSASTPGY SSLSVIEMIL AAIFFVVYMC DLHTKIPFIN WPWSDFFRTL IAAILYLITS IVVLVERGNH SKIVAGVLGL IATCLFGYDA YVTFPVRQPR HTAAPTDPAD GPV (1-152 aa encoded by BC109066)
PLP2 may play a role in cell differentiation in the intestinal epithelium.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A4 differentiation-dependent protein; Differentiation-dependent protein A4; Intestinal membrane A4 protein; Proteolipid protein 2; proteolipid protein 2 (colonic epithelium-enriched)
Gene Aliases: A4; A4LSB; PLP2
UniProt ID: (Human) Q04941
Entrez Gene ID: (Human) 5355
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support