Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
PLVAP involved in the formation of stomatal and fenestral diaphragms of caveolae. This protein may function in microvascular permeability.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fenestrated endothelial-linked structure protein; fenestrated-endothelial linked structure protein; PV-1 protein; Plasmalemma vesicle protein 1; Plasmalemma vesicle-associated protein; PV-1; PV-1 protein
Gene Aliases: FELS; gp68; PLVAP; PV-1; PV1
UniProt ID: (Human) Q9BX97
Entrez Gene ID: (Human) 83483, (Pig) 110259322
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support