Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MHSQVFSAFP YSLPLRFWVN VIKNPQFVFD IHKGSITDAC LSVVAQTFMD SCSTSEHRLD KDSPSNKLLY AKDIPSYKSW VERYYADIAK LPAISDQDMN AYLAEQSRLH AVEFNMLSAL NEIYSYVSKY SEELIGALEQ DEQARRQRLA YKVEQLINAM SIES
Plexins are a family of large, transmembrane receptors for multiple classes of semaphorins in vertebrates. Plexins are widely expressed, and regions of their extracellular domain are homologus to both scatter factor receptors and semaphorin domains. Plexins may act as semaphorin receptors alone or in combination with neuropilins. Plexins are divided into four subfamilies designated plexin-A, -B, -C, and -D. Drosophila plexin-A is a receptor for class I semaphorins and controls motor and axon guidance. Plexin-A3 mediates cell-repelling cues. Plexins B and C are receptors for Sema 4 and Sema 7, respectively.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ11751; FLJ30634; Plex 2; plexin 2; plexin-2; Plexin-A2; Semaphorin receptor OCT; transmembrane protein OCT
Gene Aliases: 2810428A13Rik; AA589422; AW457381; KIAA0463; mKIAA0463; OCT; PlexA2; PLXN2; PLXNA2; UNQ209/PRO235
UniProt ID: (Human) O75051, (Mouse) P70207
Entrez Gene ID: (Human) 5362, (Rat) 289392, (Mouse) 18845
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support