Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
This antibody has specificity for Human PMVK/phosphomevalonate kinase.
PMVK is a peroxisomal enzyme that catalyzes the conversion of mevalonate 5-phosphate into mevalonate 5-diphosphate as the fifth reaction of the cholesterol biosynthetic pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPMK; Phosphomevalonate kinase; PMKase; testis tissue sperm-binding protein Li 95mP
Gene Aliases: HUMPMKI; PMK; PMKA; PMKASE; PMKI; PMVK; POROK1
UniProt ID: (Human) Q15126
Entrez Gene ID: (Human) 10654
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support