Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
PMVK is a peroxisomal enzyme that catalyzes the conversion of mevalonate 5-phosphate into mevalonate 5-diphosphate as the fifth reaction of the cholesterol biosynthetic pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPMK; Phosphomevalonate kinase; PMKase; testis tissue sperm-binding protein Li 95mP
Gene Aliases: 1110011E12Rik; 2900002L22Rik; AV001327; HUMPMKI; PMK; PMKA; PMKASE; PMKI; PMVK; POROK1
UniProt ID: (Human) Q15126, (Mouse) Q9D1G2
Entrez Gene ID: (Human) 10654, (Mouse) 68603
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support