Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Positive Control: A549 cell lysate, HCT116 cell lysate, mouse pancreas tissue lysate, A549, SiHa.
Subcellular Location: Secreted.
This gene is a member of the lipase gene family. It encodes a carboxyl esterase that hydrolyzes insoluble, emulsified triglycerides, and is essential for the efficient digestion of dietary fats. This gene is expressed specifically in the pancreas.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Pancreatic triacylglycerol lipase; pancreatic triglyceride lipase; PL; RP11-33D13.1
Gene Aliases: 1810007A24Rik; PANLI; PL; PNLIP; PNLIPD; PTL
UniProt ID: (Human) P16233, (Rat) P27657, (Mouse) Q6P8U6
Entrez Gene ID: (Human) 5406, (Rat) 25702, (Mouse) 69060
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support