Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSGADRSPN AGAAPDSAPG QAAVASAYQR FEPRAYLRNN YAPPRGDLCN PNGVGPWKLR CLAQTFATGE VSGRTLIDIG SGPTVYQLLS ACSHFEDITM TDFLEVNRQE LGRWLQEEPG AFNWSMYSQH ACLIEGKGEC WQDKERQLRA RVKRVLPIDV HQPQPLGAGS PAPLPADALV SAFCLEAVSP DLASFQRALD HITTLLRPGG HLLLIGALEE SWYLAGEARL TVVPVSEEEV REALVRSGYK VRDLRTYIMP AHLQTGVDDV KGVFFAWAQK VGL (1-282 aa encoded by BC037246)
The product of this gene catalyzes the last step of the catecholamine biosynthesis pathway, which methylates norepinephrine to form epinephrine (adrenaline). The enzyme also has beta-carboline 2N-methyltransferase activity. This gene is thought to play a key step in regulating epinephrine production.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Noradrenaline N-methyltransferase; phenylethanolamine N-methylase; Phenylethanolamine N-methyltransferase; PNMTase
Gene Aliases: PENT; PNMT; PNMTase
UniProt ID: (Human) P11086, (Mouse) P40935, (Rat) P10937
Entrez Gene ID: (Human) 5409, (Mouse) 18948, (Rat) 24661
Molecular Function: methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support