Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEEDQELER KAIEELLKEA KRGKTRAETM GPMGWMKCPL ASTNKRFLIN TIKNTLPSHK EQDHEQKEGD KEPAKSQAQK EENPKKHRSH PYKHSFRARG SASYSPPRKR SSQDKYEKRS NRR (1-122 aa encoded by BC015319)
The protein encoded by this gene is a component of the RNA polymerase I and RNA polymerase III complexes, which function in the synthesis of ribosomal RNA precursors and small RNAs, respectively. Mutations in this gene are a cause of Treacher Collins syndrome (TCS), a craniofacial development disorder. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AC19; DNA-directed RNA polymerase I 16 kDa polypeptide; DNA-directed RNA polymerase I subunit D; DNA-directed RNA polymerases I and III subunit RPAC2; hRPA19; polymerase (RNA) I polypeptide D; polymerase (RNA) I polypeptide D, 16kDa; polymerase (RNA) I subunit D; Protein POLR1D, isoform 2; RNA polymerase 1-3; RNA polymerase I 16 kDa subunit; RNA polymerases I and III subunit AC2; RPA16; RPC16
Gene Aliases: 1110003G10Rik; AC19; POLR1C; POLR1D; RPA16; RPA9; RPAC2; RPC16; RPO1-3; TCS2
UniProt ID: (Human) P0DPB5, (Mouse) P97304
Entrez Gene ID: (Human) 51082, (Mouse) 20018, (Rat) 360762
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support