Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAGGSDPR AGDVEEDASQ LIFPKEFETA ETLLNSEVHM LLEHRKQQNE SAEDEQELSE VFMKTLNYTA RFSRFKNRET IASVRSLLLQ KKLHKFELAC LANLCPETAE ESKALIPSLE GRFEDEELQQ ILDDIQTKRS FQY (1-142 aa encoded by BC017205)
This gene encodes the fourth largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. In yeast, this polymerase subunit is associated with the polymerase under suboptimal growth conditions and may have a stress protective role. A sequence for a ribosomal pseudogene is contained within the 3' untranslated region of the transcript from this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA-directed RNA polymerase II 16 kDa polypeptide; DNA-directed RNA polymerase II subunit D; DNA-directed RNA polymerase II subunit RPB4; plasma; polymerase (RNA) II (DNA directed) polypeptide D; polymerase (RNA) II subunit D; RNA polymerase II 16 kDa subunit; RNA polymerase II subunit B4; RNA polymerase II subunit hsRBP4; RPB16
Gene Aliases: 2310002G05Rik; 2610028L19Rik; HSRBP4; HSRPB4; POLR2D; RBP4; RPB16
UniProt ID: (Human) O15514, (Mouse) Q9D7M8
Entrez Gene ID: (Human) 5433, (Mouse) 69241, (Rat) 364834
Molecular Function:
DNA-directed RNA polymerase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support