Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MNAPPAFESF LLFEGEKKIT INKDTKVPNA CLFTINKEDH TLGNIIKSQL LKDPQVLFAG YKVPHPLEHK IIIRVQTTPD YSPQEAFTNA ITDLISELSL LEERFRVAIK DKQEGIE
DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase II which synthesizes mRNA precursors and many functional non-coding RNAs. Pol II is the central component of the basal RNA polymerase II transcription machinery. It is composed of mobile elements that move relative to each other. RPB11 is part of the core element with the central large cleft.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA-directed RNA polymerase II subunit J-1; DNA-directed RNA polymerase II subunit RPB11-a; polymerase (RNA) II (DNA directed) polypeptide J, 13.3kDa; polymerase (RNA) II subunit J; RNA polymerase II 13.3 kDa subunit; RNA polymerase II subunit B11-a
Gene Aliases: hRPB14; POLR2J; POLR2J1; RPB11; RPB11A; RPB11m
UniProt ID: (Human) P52435
Entrez Gene ID: (Human) 5439
Molecular Function:
DNA-directed RNA polymerase
RNA metabolism protein
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support