Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
RPC8 is DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Specific peripheric component of RNA polymerase III which synthesizes small RNAs, such as 5S rRNA and tRNAs. It is plays a key role in sensing and limiting infection by intracellular bacteria and DNA viruses. It is acting as nuclear and cytosolic DNA sensor involved in innate immune response. It can sense non-self dsDNA that serves as template for transcription into dsRNA. The non-self RNA polymerase III transcripts, such as Epstein-Barr virus-encoded RNAs (EBERs) induce type I interferon and NF-Kappa-B through the RIG-I pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA-directed RNA polymerase III subunit 22.9 kDa polypeptide; DNA-directed RNA polymerase III subunit H; DNA-directed RNA polymerase III subunit RPC8; polymerase (RNA) III (DNA directed) polypeptide H (22.9kD); polymerase (RNA) III subunit H; RNA nucleotidyltransferase (DNA-directed); RNA polymerase III subunit 22.9 kDa subunit; RNA polymerase III subunit C7; RNA polymerase III subunit C8; RPC22.9
Gene Aliases: 5031409G22Rik; KIAA1665; POLR3H; RPC22.9; RPC8
UniProt ID: (Human) Q9Y535, (Mouse) Q9D2C6
Entrez Gene ID: (Human) 171568, (Mouse) 78929, (Rat) 300088
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support