Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MVCSPVTLRI APPDRRFSRS AIPEQIISST LSSPSSNAPD PCAKETVLSA LKEKKKK
POM121 (POM121 membrane glycoprotein), also known as POM121A (nuclear envelope pore membrane protein POM 121A), NUP121 (nucleoporin Nup121) or pore membrane protein of 121 kDa, is a 1,249 amino acid single-pass membrane protein that exists as three alternatively spiced isoforms and belongs to the POM121 family. Encoded by a gene that maps to human chromosome 7q11.23, POM121 is highly conserved and localizes to nucleus membrane and endoplasmic reticulum membrane. An essential component of the nuclear pore complex (NPC), POM121 is mandatory for nuclear envelope formation and may play a role in biogenesis of the NPC. POM121 associates with the central spoke ring complex and is involved in anchoring components of the pore complex to the pore membrane by way of its F-X-F-G repeat-containing domain. POM121 overexpression induces the formation of cytoplasmic annulate lamellae (AL).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nuclear envelope pore membrane protein POM 121; Nuclear envelope pore membrane protein POM 121A; nuclear pore membrane protein 121 kDa; nuclear pore membrane protein 121-2; Nucleoporin Nup121; POM121 membrane glycoprotein; POM121 membrane glycoprotein C; POM121B; Pore membrane protein of 121 kDa; pore membrane protein of 121 kDa C
Gene Aliases: KIAA0618; NUP121; P145; POM121; POM121-2; POM121A
UniProt ID: (Human) Q96HA1
Entrez Gene ID: (Human) 9883, (Human) 100101267
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support