Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: GDGFFSSAFQ ARLSGNNLHN ASIPEHLAYG SVITVKNLRM AIGYLHSHRH LYPEGIGARQ QQVTTYLHKD YNNLWIIKKH NTNSDPLDPS FPVEFVRHGD IIRLEHKETS RNLHSHYHEA PMTRKHYQVT GYGINGT
Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
The protein encoded by this gene is an O-mannosyltransferase that requires interaction with the product of the POMT1 gene for enzymatic function. The encoded protein is found in the membrane of the endoplasmic reticulum. Defects in this gene are a cause of Walker-Warburg syndrome (WWS).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686G10254; Dolichyl-phosphate-mannose--protein mannosyltransferase; Dolichyl-phosphate-mannose--protein mannosyltransferase 2; FLJ22309; Protein O-mannosyl-transferase 2; protein-O-mannosyltransferase 2
Gene Aliases: A830009D15Rik; AW046274; LGMD2N; MDDGA2; MDDGB2; MDDGC2; POMT2
UniProt ID: (Human) Q9UKY4, (Mouse) Q8BGQ4
Entrez Gene ID: (Human) 29954, (Rat) 688673, (Mouse) 217734
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support