Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGRLVAVGL LGIALALLGE RLLALRNRLK ASREVESVDL PHCHLIKGIE AGSEDIDILP NGLAFFSVGL KFPGLHSFAP DKPGGILMMD LKEEKPRARE LRISRGFDLA SFNPHGISTF IDNDDTVYLF VVNHPEFKNT VEIFKFEEGE NSLLHLKTVK HELLPSVNDI TAVGPAHFYA TNDHYFSDPF LKYLETYLNL HWANVVYYSP NEVKVVYLCC (1-219 aa encoded by BC046160)
This gene encodes a member of the paraoxonase gene family, which includes three known members located adjacent to each other on the long arm of chromosome 7. The encoded protein is ubiquitously expressed in human tissues, membrane-bound, and may act as a cellular antioxidant, protecting cells from oxidative stress. Hydrolytic activity against acylhomoserine lactones, important bacterial quorum-sensing mediators, suggests the encoded protein may also play a role in defense responses to pathogenic bacteria. Mutations in this gene may be associated with vascular disease and a number of quantitative phenotypes related to diabetes. Alternatively spliced transcript variants encoding different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A esterase 2; A-esterase 2; Aromatic esterase 2; arylesterase 2; paraoxonase nirs; PON 2; Serum aryldialkylphosphatase 2; Serum paraoxonase/arylesterase 2
Gene Aliases: PON2
UniProt ID: (Human) Q15165
Entrez Gene ID: (Human) 5445
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support