Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: PAALTSHPHH AVHQGLEGDL LEHISPTLSV SGLGAPEHSV MPAQIHPHHL GAMGHLHQAM GMSHPHTVAP HSAMPACLSD VESDPRELEA F
POU4F3 encodes a member of the POU-domain family of transcription factors. POU-domain proteins have been observed to play important roles in control of cell identity in several systems. This protein is found in the retina and may play a role in determining or maintaining the identities of a small subset of visual system neurons. Defects in this gene are the cause of non-syndromic sensorineural deafness autosomal dominant type 15.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain-3C; Brain-specific homeobox/POU domain protein 3C; brn-3C; MGC138412; POU domain, class 4, transcription factor 3
Gene Aliases: BRN3C; DFNA15; POU4F3
UniProt ID: (Human) Q15319
Entrez Gene ID: (Human) 5459
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support