Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 6
Purity is > 95% by SDS-PAGE.
The protein encoded by this gene is one of the three catalytic subunits of protein phosphatase 1 (PP1). PP1 is a serine/threonine specific protein phosphatase known to be involved in the regulation of a variety of cellular processes, such as cell division, glycogen metabolism, muscle contractility, protein synthesis, and HIV-1 viral transcription. Mouse studies suggest that PP1 functions as a suppressor of learning and memory. Two alternatively spliced transcript variants encoding distinct isoforms have been observed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory sperm binding protein Li 80p; MGC21657; PP-1B; PP2C-beta-X; PP2CB; PP2CBETA; PPC2BETAX; protein phosphatase 1, catalytic subunit, beta isoform; protein phosphatase 1, catalytic subunit, beta isozyme; protein phosphatase 1, catalytic subunit, delta isoform; protein phosphatase 1-beta; protein phosphatase 1-delta; protein phosphatase-1a; serine/threonine protein phosphatase PP1-beta catalytic subunit; Serine/threonine-protein phosphatase PP1-beta catalytic subunit
Gene Aliases: 1200010B19; HEL-S-80p; PP-1B; PP1B; PP1beta; PPP1CB; PPP1CD
UniProt ID: (Human) P62140, (Mouse) P62141, (Rat) P62142
Entrez Gene ID: (Human) 5500, (Mouse) 19046, (Rat) 25594
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support